siRNA Details
| Gene Symbol: Itga6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053725.1 | NP_446177.1 | 11 | 1732 | ||
| XM_006234351.1 | XP_006234413.1 | 11 | 1929 | |||
| GenBank mRNA | AJ312934.1 | - | 9 | 1242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga6| Gene Symbol: Itga6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053725.1 | NP_446177.1 | 11 | 1732 | ||
| XM_006234351.1 | XP_006234413.1 | 11 | 1929 | |||
| GenBank mRNA | AJ312934.1 | - | 9 | 1242 | ||