siRNA Details
| Gene Symbol: Nfil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053727.2 | NP_446179.2 | 2 | 777 | ||
| XM_006253667.2 | XP_006253729.1 | 2 | 1451 | |||
| XM_006253668.2 | XP_006253730.1 | 2 | 832 | |||
| XM_006253669.2 | XP_006253731.1 | 2 | 2014 | |||
| XM_006253670.2 | XP_006253732.1 | 2 | 1186 | |||
| GenBank mRNA | AY004663.1 | - | 1 | 613 | ||
| BC072527.1 | - | 2 | 777 | 7097417 | ||

Nfil3
Genomic Map