siRNA Details
| Gene Symbol: Strn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240075.2 | XP_006240137.1 | 13, 14 | 2066 | ||
| XM_006240076.2 | XP_006240138.1 | 13, 14 | 2035 | |||
| XM_006240077.2 | XP_006240139.1 | 12, 13 | 1913 | |||
| GenBank mRNA | FQ222839.1 | - | 8, 9 | 1072 | ||

Strn3
Genomic Map