siRNA Details
| Gene Symbol: Stag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053730.1 | NP_446182.1 | 19 | 2114 | ||
| XM_006249020.2 | XP_006249082.1 | 19 | 2282 | |||
| GenBank mRNA | AY027880.1 | - | 19 | 2114 | ||
| Gene Symbol: LOC102553065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257725.2 | XP_006257787.2 | 3 | 348 | ||

Stag3
Genomic Map