siRNA Details
| Gene Symbol: Ntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053731.2 | NP_446183.2 | 6 | 1574 | ||
| XM_006246567.2 | XP_006246629.1 | 7 | 1926 | |||
| XM_006246568.2 | XP_006246630.1 | 7 | 1760 | |||
| GenBank mRNA | AY028417.1 | - | 6 | 1574 | ||

Ntn1
Genomic Map