siRNA Details
Gene Symbol: Bcl2l10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053733.1 | NP_446185.1 | 1 | 370 | ||
GenBank mRNA | AY029163.1 | - | 1 | 370 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bcl2l10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053733.1 | NP_446185.1 | 1 | 370 | ||
GenBank mRNA | AY029163.1 | - | 1 | 370 |