siRNA Details
| Gene Symbol: Pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053735.1 | NP_446187.1 | 8 | 1316 | ||
| GenBank mRNA | AY029199.1 | - | 8 | 1316 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pi4k2a| Gene Symbol: Pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053735.1 | NP_446187.1 | 8 | 1316 | ||
| GenBank mRNA | AY029199.1 | - | 8 | 1316 | ||