siRNA Details
| Gene Symbol: Arhgef7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113521.1 | NP_001106993.1 | 13, 14 | 1473 | ||
| NM_001113522.1 | NP_001106994.1 | 13, 14 | 1473 | |||
| NM_053740.2 | NP_446192.2 | 10, 11 | 1107 | |||
| XM_008771376.1 | XP_008769598.1 | 13, 14 | 12445 | |||
| XM_008771377.1 | XP_008769599.1 | 13, 14 | 12444 | |||
| XM_008771378.1 | XP_008769600.1 | 13, 14 | 12445 | |||
| XM_008771379.1 | XP_008769601.1 | 1530 | ||||
| XM_008771380.1 | XP_008769602.1 | 1443 | ||||
| GenBank mRNA | AF044673.1 | - | 10, 11 | 1107 | ||
| AY034823.1 | - | 10, 11 | 1107 | |||
| AY996221.1 | - | 10, 11 | 1107 | |||
| BC131844.1 | - | 11, 12 | 1160 | |||

Arhgef7
Genomic Map