siRNA Details
| Gene Symbol: Pitpnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053742.1 | NP_446194.1 | 10 | 736 | ||
| XM_006249522.2 | XP_006249584.1 | 10 | 780 | |||
| GenBank mRNA | BC061538.1 | - | 10 | 768 | 6888321 | |
| D21132.1 | - | 10 | 736 | |||

Pitpnb
Genomic Map