siRNA Details
| Gene Symbol: Ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053747.2 | NP_446199.2 | 8, 9 | 1472 | ||
| GenBank mRNA | BC072477.1 | - | 8, 9 | 1472 | 7103662 | |
| D87950.1 | - | 8, 9 | 1332 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubqln1| Gene Symbol: Ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053747.2 | NP_446199.2 | 8, 9 | 1472 | ||
| GenBank mRNA | BC072477.1 | - | 8, 9 | 1472 | 7103662 | |
| D87950.1 | - | 8, 9 | 1332 | |||