siRNA Details
| Gene Symbol: Nppc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053750.1 | NP_446202.1 | 3 | 828 | ||
| GenBank mRNA | D90219.1 | - | 3 | 828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nppc| Gene Symbol: Nppc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053750.1 | NP_446202.1 | 3 | 828 | ||
| GenBank mRNA | D90219.1 | - | 3 | 828 | ||