siRNA Details
| Gene Symbol: Wap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053751.4 | NP_446203.1 | 3 | 375 | ||
| XM_008770244.1 | XP_008768466.1 | 3 | 742 | |||
| GenBank mRNA | J00802.1 | - | 3 | 375 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wap| Gene Symbol: Wap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053751.4 | NP_446203.1 | 3 | 375 | ||
| XM_008770244.1 | XP_008768466.1 | 3 | 742 | |||
| GenBank mRNA | J00802.1 | - | 3 | 375 | ||