siRNA Details
| Gene Symbol: Abcg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053754.2 | NP_446206.2 | 8 | 1134 | ||
| XM_008764452.1 | XP_008762674.1 | 9 | 1374 | |||
| GenBank mRNA | AF312714.3 | - | 8 | 1134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcg5| Gene Symbol: Abcg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053754.2 | NP_446206.2 | 8 | 1134 | ||
| XM_008764452.1 | XP_008762674.1 | 9 | 1374 | |||
| GenBank mRNA | AF312714.3 | - | 8 | 1134 | ||