siRNA Details
| Gene Symbol: Zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053761.1 | NP_446213.1 | 9 | 1723 | ||
| XM_006236387.2 | XP_006236449.1 | 9 | 1878 | |||
| XM_006236388.2 | XP_006236450.1 | 9 | 1841 | |||
| XM_006236389.2 | XP_006236451.1 | 8 | 1785 | |||
| GenBank mRNA | AF329827.1 | - | 2 | 167 | ||
| FQ234073.1 | - | 3 | 320 | |||

Zyx
Genomic Map