siRNA Details
| Gene Symbol: Mapre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138509.3 | NP_612518.2 | 4 | 394 | ||
| XM_006235294.2 | XP_006235356.1 | 4 | 377 | |||
| XM_006235295.2 | XP_006235357.1 | 4 | 447 | |||
| GenBank mRNA | BC081726.1 | - | 4 | 394 | 7192469 | |

Mapre1
Genomic Map