siRNA Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053766.1 | NP_446218.1 | 7 | 766 | ||
| XM_008770840.1 | XP_008769062.1 | 4 | 1282 | |||
| XM_008770841.1 | XP_008769063.1 | 5 | 785 | |||
| GenBank mRNA | BC089052.1 | - | 7 | 766 | 7307299 | |
| FQ226684.1 | - | 6 | 548 | |||

Wbp4
Genomic Map