siRNA Details
| Gene Symbol: Ptpre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053767.1 | NP_446219.1 | 6 | 579 | ||
| XM_006230449.2 | XP_006230511.1 | 8 | 3500 | |||
| XM_006230450.2 | XP_006230512.1 | 5 | 507 | |||
| XM_006230451.2 | XP_006230513.1 | 6 | 2811 | |||
| XM_008759971.1 | XP_008758193.1 | 7 | 1178 | |||
| XM_008759972.1 | XP_008758194.1 | 7 | 1679 | |||
| GenBank mRNA | BC166573.1 | - | 6 | 589 | 7320594 | |
| D78610.1 | - | 5 | 453 | |||
| D78613.1 | - | 6 | 572 | |||

Ptpre
Genomic Map