siRNA Details
| Gene Symbol: Ptpre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053767.1 | NP_446219.1 | 5 | 458 | ||
| XM_006230449.2 | XP_006230511.1 | 7 | 3379 | |||
| XM_006230450.2 | XP_006230512.1 | 4 | 386 | |||
| XM_006230451.2 | XP_006230513.1 | 5 | 2690 | |||
| XM_008759971.1 | XP_008758193.1 | 6 | 1057 | |||
| XM_008759972.1 | XP_008758194.1 | 6 | 1558 | |||
| GenBank mRNA | BC166573.1 | - | 5 | 468 | 7320594 | |
| D78610.1 | - | 4 | 332 | |||
| D78613.1 | - | 5 | 451 | |||

Ptpre
Genomic Map