siRNA Details
| Gene Symbol: Ccnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100472.1 | NP_001093942.1 | 7, 8 | 558 | ||
| XM_006237937.2 | XP_006237999.1 | 7, 8 | 642 | |||
| GenBank mRNA | BC161908.1 | - | 8, 9 | 678 | ||
| D14013.1 | - | 7, 8 | 543 | |||

Ccnc
Genomic Map