siRNA Details
| Gene Symbol: Cdkn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080782.3 | NP_542960.1 | 2 | 453 | ||
| XM_006256128.2 | XP_006256190.1 | 2 | 517 | |||
| GenBank mRNA | BC100620.1 | - | 2 | 453 | 7938247 | |
| U24174.1 | - | 2 | 496 | |||

Cdkn1a
Genomic Map