siRNA Details
| Gene Symbol: Dusp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053769.3 | NP_446221.2 | 3 | 710 | ||
| GenBank mRNA | S81478.1 | - | 3 | 684 | ||
| U02553.1 | - | 3 | 696 | |||
| X84004.1 | - | 3 | 733 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp1| Gene Symbol: Dusp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053769.3 | NP_446221.2 | 3 | 710 | ||
| GenBank mRNA | S81478.1 | - | 3 | 684 | ||
| U02553.1 | - | 3 | 696 | |||
| X84004.1 | - | 3 | 733 | |||