siRNA Details
| Gene Symbol: Shc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105743.1 | NP_001099213.1 | 2, 3 | 538 | ||
| GenBank mRNA | AB001453.1 | - | 2, 3 | 752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shc3| Gene Symbol: Shc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105743.1 | NP_001099213.1 | 2, 3 | 538 | ||
| GenBank mRNA | AB001453.1 | - | 2, 3 | 752 | ||