siRNA Details
| Gene Symbol: TARBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005646.3 | NP_005637.3 | 20 | 3425 | ||
| XM_005273234.3 | XP_005273291.1 | 20 | 3422 | |||
| XM_011544263.2 | XP_011542565.1 | 19 | 3420 | |||
| XM_017002194.1 | XP_016857683.1 | 20 | 3495 | |||
| XM_017002195.1 | XP_016857684.1 | 17 | 2298 | |||
| XM_017002196.1 | XP_016857685.1 | 8 | 1270 | |||
| XM_017002197.1 | XP_016857686.1 | 15 | 2038 | |||
| GenBank mRNA | U38847.1 | - | 20 | 3425 | ||

TARBP1
Genomic Map