siRNA Details
| Gene Symbol: Cacna1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153814.2 | NP_722521.1 | 26 | 5005 | ||
| XM_008767532.1 | XP_008765754.1 | 26 | 9402 | |||
| XM_008767533.1 | XP_008765755.1 | 26 | 9402 | |||
| XM_008767534.1 | XP_008765756.1 | 25 | 9384 | |||
| XM_008767535.1 | XP_008765757.1 | 25 | 9384 | |||
| XM_008767536.1 | XP_008765758.1 | 25 | 4621 | |||
| GenBank mRNA | AF290213.2 | - | 26 | 5008 | ||

Cacna1h
Genomic Map