siRNA Details
| Gene Symbol: TARBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005646.3 | NP_005637.3 | 29 | 4654 | ||
| XM_005273234.3 | XP_005273291.1 | 29 | 4651 | |||
| XM_011544263.2 | XP_011542565.1 | 28 | 4649 | |||
| XM_017002195.1 | XP_016857684.1 | 26 | 3527 | |||
| XM_017002196.1 | XP_016857685.1 | 17 | 2499 | |||
| XM_017002197.1 | XP_016857686.1 | 24 | 3267 | |||
| GenBank mRNA | AK055044.1 | - | 6 | 2636 | ||
| BC016023.1 | - | 7 | 1243 | |||
| U38847.1 | - | 29 | 4654 | |||

TARBP1
Genomic Map