siRNA Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053773.1 | NP_446225.1 | 14 | 2071 | ||
| XM_006231175.2 | XP_006231237.1 | 14 | 3153 | |||
| XM_006231176.2 | XP_006231238.1 | 14 | 3154 | |||
| XM_006231177.2 | XP_006231239.1 | 13 | 2384 | |||
| XM_006231178.2 | XP_006231240.1 | 13 | 5313 | |||
| XM_006231179.2 | XP_006231241.1 | 13 | 2217 | |||
| XM_006231181.2 | XP_006231243.1 | 14 | 2172 | |||
| XM_006231182.2 | XP_006231244.1 | 13 | 2384 | |||
| XM_008760302.1 | XP_008758524.1 | 14 | 1913 | |||
| GenBank mRNA | BC103479.1 | - | 14 | 2071 | 7936785 | |
| U75916.1 | - | 7 | 704 | |||

Tjp2
Genomic Map