siRNA Details
| Gene Symbol: Usp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053774.2 | NP_446226.2 | 2 | 741 | ||
| GenBank mRNA | AF106659.1 | - | 2 | 741 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp2| Gene Symbol: Usp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053774.2 | NP_446226.2 | 2 | 741 | ||
| GenBank mRNA | AF106659.1 | - | 2 | 741 | ||