siRNA Details
| Gene Symbol: Usp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053774.2 | NP_446226.2 | 11 | 1709 | ||
| XM_006242861.2 | XP_006242923.1 | 11 | 1870 | |||
| XM_006242863.2 | XP_006242925.1 | 10 | 1161 | |||
| XM_008766138.1 | XP_008764360.1 | 10 | 1010 | |||
| XM_008766139.1 | XP_008764361.1 | 10 | 1013 | |||
| GenBank mRNA | AF106658.1 | - | 10 | 1137 | ||
| AF106659.1 | - | 11 | 1709 | |||
| AF202453.1 | - | 10 | 1582 | |||
| AF202454.1 | - | 10 | 916 | |||
| BC085719.1 | - | 11 | 1683 | 7193298 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_016808, NM_198092, NM_198091, AK138799, BC017517, and others.

Usp2
Genomic Map