siRNA Details
| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053775.3 | NP_446227.2 | 8 | 803 | ||
| GenBank mRNA | BC168659.1 | - | 888 | 7938299 | ||
| DQ286425.1 | - | 8 | 803 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v0a2| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053775.3 | NP_446227.2 | 8 | 803 | ||
| GenBank mRNA | BC168659.1 | - | 888 | 7938299 | ||
| DQ286425.1 | - | 8 | 803 | |||