siRNA Details
| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053775.3 | NP_446227.2 | 19, 20 | 2463 | ||
| GenBank mRNA | BC098791.1 | - | 1324 | |||
| BC168659.1 | - | 2548 | 7938299 | |||
| DQ286425.1 | - | 19, 20 | 2463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v0a2| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053775.3 | NP_446227.2 | 19, 20 | 2463 | ||
| GenBank mRNA | BC098791.1 | - | 1324 | |||
| BC168659.1 | - | 2548 | 7938299 | |||
| DQ286425.1 | - | 19, 20 | 2463 | |||