siRNA Details
| Gene Symbol: Dnajc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053776.2 | NP_446228.2 | 2, 3 | 403 | ||
| XM_008762628.1 | XP_008760850.1 | 3, 4 | 712 | |||
| GenBank mRNA | AF118853.1 | - | 2, 3 | 376 | ||
| AY322161.1 | - | 2, 3 | 376 | |||
| AY322162.1 | - | 2, 3 | 376 | |||
| AY322163.1 | - | 2, 3 | 287 | |||
| AY322164.1 | - | 2, 3 | 287 | |||
| BC088838.1 | - | 2, 3 | 403 | 7132881 | ||

Dnajc2
Genomic Map