siRNA Details
| Gene Symbol: Ncoa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276714.1 | NP_001263643.1 | 9 | 4886 | ||
| XM_006235296.2 | XP_006235358.1 | 11 | 5223 | |||
| XM_006235297.1 | XP_006235359.1 | 11 | 5120 | |||
| XM_008762295.1 | XP_008760517.1 | 12 | 5292 | |||
| XM_008762296.1 | XP_008760518.1 | 11 | 5120 | |||
| XM_008762297.1 | XP_008760519.1 | 11 | 5087 | |||
| XM_008762298.1 | XP_008760520.1 | 11 | 5084 | |||

Ncoa6
Genomic Map