siRNA Details
| Gene Symbol: Ncoa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276714.1 | NP_001263643.1 | 7 | 2647 | ||
| XM_006235296.2 | XP_006235358.1 | 9 | 2984 | |||
| XM_006235297.1 | XP_006235359.1 | 9 | 2881 | |||
| XM_008762295.1 | XP_008760517.1 | 10 | 3053 | |||
| XM_008762296.1 | XP_008760518.1 | 9 | 2881 | |||
| XM_008762297.1 | XP_008760519.1 | 9 | 2848 | |||
| XM_008762298.1 | XP_008760520.1 | 9 | 2845 | |||
| XM_008762299.1 | XP_008760521.1 | 9 | 2881 | |||
| GenBank mRNA | AF176351.1 | - | 1 | 520 | ||
| AF228043.1 | - | 1 | 118 | |||

Ncoa6
Genomic Map