siRNA Details
Gene Symbol: Ifngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053783.1 | NP_446235.1 | 3 | 310 | ||
GenBank mRNA | AF201901.1 | - | 3 | 310 | ||
BC062039.1 | - | 3 | 252 | 5598500 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053783.1 | NP_446235.1 | 3 | 310 | ||
GenBank mRNA | AF201901.1 | - | 3 | 310 | ||
BC062039.1 | - | 3 | 252 | 5598500 |