siRNA Details
| Gene Symbol: Stx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053788.2 | NP_446240.2 | 4 | 268 | ||
| GenBank mRNA | AF217191.1 | - | 4 | 237 | ||
| D10392.1 | - | 4 | 268 | |||
| D12519.1 | - | 4 | 228 | |||
| M95734.1 | - | 4 | 229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stx1a| Gene Symbol: Stx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053788.2 | NP_446240.2 | 4 | 268 | ||
| GenBank mRNA | AF217191.1 | - | 4 | 237 | ||
| D10392.1 | - | 4 | 268 | |||
| D12519.1 | - | 4 | 228 | |||
| M95734.1 | - | 4 | 229 | |||