siRNA Details
Gene Symbol: Il17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053789.1 | NP_446241.1 | 2 | 317 | ||
GenBank mRNA | AF218724.1 | - | 1 | 87 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053789.1 | NP_446241.1 | 2 | 317 | ||
GenBank mRNA | AF218724.1 | - | 1 | 87 |