siRNA Details
| Gene Symbol: Prl2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053791.2 | NP_446243.1 | 4 | 457 | ||
| GenBank mRNA | AF226608.1 | - | 4 | 455 | ||
| BC097296.1 | - | 4 | 457 | 7445776 | ||
| DQ329279.1 | - | 4 | 455 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl2a1| Gene Symbol: Prl2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053791.2 | NP_446243.1 | 4 | 457 | ||
| GenBank mRNA | AF226608.1 | - | 4 | 455 | ||
| BC097296.1 | - | 4 | 457 | 7445776 | ||
| DQ329279.1 | - | 4 | 455 | |||