siRNA Details
| Gene Symbol: Ift172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053792.1 | NP_446244.1 | 41 | 4544 | ||
| GenBank mRNA | AF226993.1 | - | 41 | 4544 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026298, BC066096, AK122461, BC051928, AY339616.

Ift172
Genomic Map