siRNA Details
| Gene Symbol: Kidins220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053795.1 | NP_446247.1 | 30 | 5020 | ||
| XM_006239923.2 | XP_006239985.1 | 31 | 5524 | |||
| XM_006239924.2 | XP_006239986.1 | 31 | 5521 | |||
| XM_006239925.2 | XP_006239987.1 | 32 | 5214 | |||
| XM_006239926.2 | XP_006239988.1 | 31 | 5509 | |||
| XM_006239927.2 | XP_006239989.1 | 32 | 5211 | |||
| XM_006239928.2 | XP_006239990.1 | 32 | 5196 | |||
| XM_006239929.2 | XP_006239991.1 | 30 | 5491 | |||
| XM_006239931.2 | XP_006239993.1 | 30 | 5467 | |||
| XM_006239932.2 | XP_006239994.1 | 30 | 5452 | |||
| XM_006239933.2 | XP_006239995.1 | 29 | 5433 | |||
| XM_006239934.2 | XP_006239996.1 | 29 | 5418 | |||
| XM_006239935.2 | XP_006239997.1 | 28 | 5385 | |||
| XM_006239936.2 | XP_006239998.1 | 27 | 5271 | |||
| XM_006239937.2 | XP_006239999.1 | 28 | 4962 | |||
| GenBank mRNA | AF239045.2 | - | 30 | 5020 | ||
| AF313464.1 | - | 27 | 4768 | |||

Kidins220
Genomic Map