siRNA Details
| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053798.2 | NP_446250.1 | 8 | 642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sacm1l| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053798.2 | NP_446250.1 | 8 | 642 | ||