siRNA Details
| Gene Symbol: Txn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053800.3 | NP_446252.1 | 3 | 204 | ||
| GenBank mRNA | AF311055.1 | - | 3 | 204 | ||
| BC058454.1 | - | 289 | 6886988 | |||
| FQ209943.1 | - | 286 | ||||
| FQ210839.1 | - | 289 | ||||
| FQ216849.1 | - | 315 | ||||
| FQ217822.1 | - | 284 | ||||
| FQ221646.1 | - | 257 | ||||
| X14878.1 | - | 264 | ||||

Txn1
Genomic Map