siRNA Details
| Gene Symbol: Tgfbi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053802.1 | NP_446254.1 | 5, 6 | 658 | ||
| GenBank mRNA | CD373288.1 | - | 2, 3 | 230 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgfbi| Gene Symbol: Tgfbi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053802.1 | NP_446254.1 | 5, 6 | 658 | ||
| GenBank mRNA | CD373288.1 | - | 2, 3 | 230 | ||