siRNA Details
| Gene Symbol: Kcnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053804.2 | NP_446256.2 | 1 | 44 | ||
| GenBank mRNA | AF302842.2 | - | 1 | 44 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk4| Gene Symbol: Kcnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053804.2 | NP_446256.2 | 1 | 44 | ||
| GenBank mRNA | AF302842.2 | - | 1 | 44 | ||