siRNA Details
| Gene Symbol: Gripap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053807.1 | NP_446259.1 | 3 | 202 | ||
| GenBank mRNA | AF274057.1 | - | 3 | 202 | ||
| AF274058.1 | - | 3 | 202 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gripap1| Gene Symbol: Gripap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053807.1 | NP_446259.1 | 3 | 202 | ||
| GenBank mRNA | AF274057.1 | - | 3 | 202 | ||
| AF274058.1 | - | 3 | 202 | |||