siRNA Details
Gene Symbol: F2rl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053808.1 | NP_446260.1 | 1 | 52 | ||
GenBank mRNA | AF269246.2 | - | 1 | 52 | ||
AF310216.1 | - | 1 | 79 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F2rl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053808.1 | NP_446260.1 | 1 | 52 | ||
GenBank mRNA | AF269246.2 | - | 1 | 52 | ||
AF310216.1 | - | 1 | 79 |