siRNA Details
| Gene Symbol: Fgf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053809.1 | NP_446261.1 | 2, 3 | 406 | ||
| XM_006230640.2 | XP_006230702.1 | 2, 3 | 536 | |||
| GenBank mRNA | AB079673.1 | - | 2, 3 | 406 | ||
| AF260830.1 | - | 1, 2 | 72 | |||

Fgf4
Genomic Map