siRNA Details
| Gene Symbol: TBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005992.1 | NP_005983.1 | 6, 7 | 976 | ||
| NM_080646.1 | NP_542377.1 | 6, 7 | 976 | |||
| NM_080647.1 | NP_542378.1 | 6, 7 | 976 | |||
| XM_006724312.2 | XP_006724375.1 | 6, 7 | 1145 | |||
| XM_017028925.1 | XP_016884414.1 | 5, 6 | 1021 | |||
| XM_017028926.1 | XP_016884415.1 | 6, 7 | 1111 | |||
| XM_017028928.1 | XP_016884417.1 | 5, 6 | 1020 | |||
| GenBank mRNA | AF012130.1 | - | 6, 7 | 976 | ||
| AF012131.1 | - | 6, 7 | 976 | |||
| AF373867.1 | - | 6, 7 | 976 | |||

TBX1
Genomic Map