siRNA Details
| Gene Symbol: CTSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001909.4 | NP_001900.1 | 1, 2 | 209 | ||
| GenBank mRNA | AK130178.1 | - | 1, 2 | 101 | ||
| BC016320.2 | - | 1, 2 | 121 | 3506977 | ||
| BM762986.1 | - | 1, 2 | 209 | |||
| BT006910.1 | - | 1, 2 | 76 | |||
| BT020155.1 | - | 1, 2 | 76 | |||
| CR456947.1 | - | 1, 2 | 76 | |||
| EU794598.1 | - | 1, 2 | 121 | |||
| KU178007.1 | - | 1, 2 | 76 | |||
| M11233.1 | - | 1, 2 | 127 | |||
| X05344.1 | - | 1, 2 | 78 | |||

CTSD
Genomic Map