siRNA Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034022.1 | NP_001029194.1 | 7, 8 | 992 | ||
| NM_053814.2 | NP_446266.2 | 7, 8 | 992 | |||
| XM_006246425.2 | XP_006246487.1 | 7, 8 | 1259 | |||
| XM_006246427.2 | XP_006246489.1 | 7, 8 | 1260 | |||
| XM_006246432.2 | XP_006246494.1 | 7, 8 | 1245 | |||
| GenBank mRNA | AF311311.1 | - | 7, 8 | 922 | ||
| BC098911.1 | - | 7, 8 | 978 | 7446349 | ||

Mprip
Genomic Map