siRNA Details
| Gene Symbol: Slc6a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053818.2 | NP_446270.1 | 13 | 1745 | ||
| XM_006238625.2 | XP_006238687.1 | 12 | 1751 | |||
| GenBank mRNA | BC128732.1 | - | 13 | 1745 | 8364347 | |
| L13600.1 | - | 12 | 1642 | |||
| M88595.1 | - | 12 | 1633 | |||
| M95413.1 | - | 13 | 1696 | |||

Slc6a9
Genomic Map